Assay Details
| Gene Symbol: mad2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244840.1 | NP_001231769.1 | 3 - 4 | 170 | 74 | |
| NM_001244841.1 | NP_001231770.1 | 2 - 3 | 106 | 74 | ||
| NM_001244842.1 | NP_001231771.1 | 2 - 3 | 130 | 74 | ||
| NR_045201.1 | 2 - 3 | 130 | 74 | |||
| XM_012966356.2 | XP_012821810.1 | 3 - 4 | 197 | 74 | ||
| XM_018095335.1 | XP_017950824.1 | 3 - 4 | 621 | 74 | ||
| XM_018095336.1 | XP_017950825.1 | 3 - 4 | 181 | 74 | ||
| XM_018095337.1 | XP_017950826.1 | 1 - 2 | 41 | 74 | ||
| GenBank mRNA | BC074654.1 | - | 2 - 3 | 119 | 74 | |
| BQ521961.1 | - | 2 - 3 | 119 | 74 | ||
| CR760993.2 | - | 2 - 3 | 118 | 74 | ||
| CX840685.1 | - | 2 - 3 | 129 | 74 | ||
| CX842973.1 | - | 1 - 2 | 55 | 74 | ||
| ES678430.1 | - | 3 - 4 | 170 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018095337.1 transcripts.
Set Membership: |
|

mad2l2
Genomic Map