Assay Details
| Gene Symbol: imp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004848.1 | NP_001004848.1 | 4 - 5 | 569 | 77 | |
| XM_018092989.1 | XP_017948478.1 | 4 - 5 | 351 | 77 | ||
| GenBank mRNA | BC074666.1 | - | 4 - 5 | 569 | 77 | |
More Information
Set Membership: |
|

imp3
Genomic Map