Assay Details
| Gene Symbol: imp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004848.1 | NP_001004848.1 | 6 - 7 | 687 | 94 | |
| XM_018092989.1 | XP_017948478.1 | 6 - 7 | 469 | 94 | ||
| GenBank mRNA | BC074666.1 | - | 6 - 7 | 687 | 94 | |
More Information
Set Membership: |
|

imp3
Genomic Map