Assay Details
| Gene Symbol: ctsv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004869.1 | NP_001004869.1 | 4 - 5 | 420 | 95 | |
| XM_012959164.1 | XP_012814618.1 | 4 - 5 | 666 | 95 | ||
| GenBank mRNA | BC074718.1 | - | 4 - 5 | 420 | 95 | |
| CR760760.2 | - | 3 - 4 | 395 | 95 | ||
More Information
Set Membership: |
|

ctsv
Genomic Map