Assay Details
| Gene Symbol: ctsv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004869.1 | NP_001004869.1 | 6 - 7 | 820 | 74 | |
| XM_012959164.1 | XP_012814618.1 | 6 - 7 | 1066 | 74 | ||
| GenBank mRNA | BC074718.1 | - | 6 - 7 | 820 | 74 | |
| CR760760.2 | - | 5 - 6 | 795 | 74 | ||
More Information
Set Membership: |
|

ctsv
Genomic Map