Assay Details
| Gene Symbol: cyp2a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004878.1 | NP_001004878.1 | 4 - 5 | 496 | 70 | |
| GenBank mRNA | BC075265.1 | - | 4 - 5 | 521 | 70 | |
More Information
Set Membership: |
|

cyp2a13
Genomic Map