Assay Details
| Gene Symbol: tmbim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004879.1 | NP_001004879.1 | 6 - 7 | 545 | 89 | |
| GenBank mRNA | BC075267.1 | - | 6 - 7 | 545 | 89 | |
| CR760482.2 | - | 6 - 7 | 507 | 89 | ||
More Information
Set Membership: |
|

tmbim4
Genomic Map