Assay Details
| Gene Symbol: tango2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004885.2 | NP_001004885.1 | 8 - 9 | 926 | 62 | |
| XM_012959343.2 | XP_012814797.1 | 9 - 10 | 1032 | 62 | ||
| XM_012959347.2 | XP_012814801.1 | 9 - 10 | 1029 | 62 | ||
| GenBank mRNA | BC075282.1 | - | 8 - 9 | 928 | 62 | |
| CR855614.2 | - | 8 - 9 | 925 | 62 | ||
More Information
Set Membership: |
|

tango2
Genomic Map