Assay Details
| Gene Symbol: btn2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004895.1 | NP_001004895.1 | 4 - 5 | 762 | 68 | |
| GenBank mRNA | BC075308.1 | - | 4 - 5 | 762 | 68 | |
More Information
Set Membership: |
|

btn2a2
Genomic Map