Assay Details
| Gene Symbol: nadk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004896.1 | NP_001004896.1 | 1 - 2 | 240 | 63 | |
| GenBank mRNA | BC075309.1 | - | 1 - 2 | 240 | 63 | |
More Information
Set Membership: |
|

nadk
Genomic Map