Assay Details
| Gene Symbol: arpin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004898.1 | NP_001004898.1 | 4 - 5 | 543 | 95 | |
| XM_012958820.2 | XP_012814274.1 | 5 - 6 | 696 | 95 | ||
| XM_012958821.2 | XP_012814275.1 | 5 - 6 | 685 | 95 | ||
| XM_012958822.2 | XP_012814276.1 | 4 - 5 | 514 | 95 | ||
| XM_012958823.2 | XP_012814277.1 | 4 - 5 | 506 | 95 | ||
| GenBank mRNA | BC075312.1 | - | 4 - 5 | 543 | 95 | |
More Information
Set Membership: |
|

arpin
Genomic Map