Assay Details
| Gene Symbol: sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004900.1 | NP_001004900.1 | 5 - 6 | 947 | 79 | |
| XM_012954910.2 | XP_012810364.1 | 5 - 6 | 2499 | 79 | ||
| GenBank mRNA | BC075315.1 | - | 5 - 6 | 947 | 79 | |
More Information
Set Membership: |
|

sult1c2
Genomic Map