Assay Details
| Gene Symbol: mitd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004902.1 | NP_001004902.1 | 6 - 7 | 712 | 76 | |
| GenBank mRNA | BC075321.1 | - | 6 - 7 | 712 | 76 | |
| CR761296.2 | - | 6 - 7 | 701 | 76 | ||
More Information
Set Membership: |
|

mitd1
Genomic Map