Assay Details
| Gene Symbol: stn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004908.1 | NP_001004908.1 | 4 - 5 | 498 | 88 | |
| XM_012966375.2 | XP_012821829.1 | 4 - 5 | 424 | 88 | ||
| GenBank mRNA | BC075336.1 | - | 4 - 5 | 498 | 88 | |
More Information
Set Membership: |
|

stn1
Genomic Map