Assay Details
| Gene Symbol: ube2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004909.1 | NP_001004909.1 | 1 - 2 | 289 | 71 | |
| GenBank mRNA | BC075340.1 | - | 1 - 2 | 289 | 71 | |
| CR761679.2 | - | 1 - 2 | 208 | 71 | ||
More Information
Set Membership: |
|

ube2h
Genomic Map