Assay Details
| Gene Symbol: med10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004914.1 | NP_001004914.1 | 1 - 2 | 245 | 86 | |
| GenBank mRNA | BC075360.1 | - | 1 - 2 | 245 | 86 | |
More Information
Set Membership: |
|

med10
Genomic Map