Assay Details
Gene Symbol: atp6v1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004928.1 | NP_001004928.1 | 1 - 2 | 231 | 108 | |
GenBank mRNA | BC075390.1 | - | 1 - 2 | 231 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
