Assay Details
Gene Symbol: bicdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004934.1 | NP_001004934.1 | 1441 | 66 | ||
GenBank mRNA | BC075409.1 | - | 1441 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
