Assay Details
| Gene Symbol: clu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004936.1 | NP_001004936.1 | 286 | 69 | ||
| GenBank mRNA | BC075303.1 | - | 286 | 69 | ||
| BC075411.1 | - | 286 | 69 | |||
| BC088762.1 | - | 288 | 69 | |||
More Information
Set Membership: |
|

clu
Genomic Map