Assay Details
| Gene Symbol: adssl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004939.1 | NP_001004939.1 | 1 - 2 | 239 | 62 | |
| GenBank mRNA | BC075419.1 | - | 1 - 2 | 239 | 62 | |
| CR848252.2 | - | 1 - 2 | 234 | 62 | ||
More Information
Set Membership: |
|

adssl1
Genomic Map