Assay Details
| Gene Symbol: rack1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004946.1 | NP_001004946.1 | 2 - 3 | 355 | 85 | |
| XM_012968220.2 | XP_012823674.1 | 3 - 4 | 479 | 85 | ||
| XM_012968221.2 | XP_012823675.1 | 3 - 4 | 605 | 85 | ||
| GenBank mRNA | BC075435.1 | - | 2 - 3 | 355 | 85 | |
| BC166955.1 | - | 2 - 3 | 347 | 85 | ||
| CR760842.1 | - | 2 - 3 | 310 | 85 | ||
| Gene Symbol: aste1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC075288.1 | - | 2875 | 85 | ||
More Information
Set Membership: |
|

rack1
Genomic Map