Assay Details
| Gene Symbol: gstm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004964.1 | NP_001004964.1 | 5 - 6 | 418 | 100 | |
| GenBank mRNA | BC075475.1 | - | 5 - 6 | 418 | 100 | |
| CR761400.2 | - | 5 - 6 | 438 | 100 | ||
More Information
Set Membership: |
|

gstm1
Genomic Map