Assay Details
| Gene Symbol: rpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004965.2 | NP_001004965.1 | 5 - 6 | 505 | 98 | |
| XM_012954243.1 | XP_012809697.1 | 6 - 7 | 544 | 98 | ||
| GenBank mRNA | BC075477.1 | - | 5 - 6 | 509 | 98 | |
| BC158397.1 | - | 5 - 6 | 519 | 98 | ||
| CU025127.1 | - | 5 - 6 | 504 | 98 | ||
More Information
Set Membership: |
|

rpl10
Genomic Map