Assay Details
| Gene Symbol: asf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004974.1 | NP_001004974.1 | 3 - 4 | 315 | 102 | |
| GenBank mRNA | BC075495.1 | - | 3 - 4 | 315 | 102 | |
| CR848421.2 | - | 3 - 4 | 351 | 102 | ||
More Information
Set Membership: |
|

asf1b
Genomic Map