Assay Details
| Gene Symbol: st13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004975.1 | NP_001004975.1 | 5 - 6 | 439 | 89 | |
| GenBank mRNA | BC075506.1 | - | 5 - 6 | 439 | 89 | |
| CR942468.2 | - | 5 - 6 | 478 | 89 | ||
More Information
Set Membership: |
|

st13
Genomic Map