Assay Details
| Gene Symbol: yeats4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004981.1 | NP_001004981.1 | 6 - 7 | 699 | 84 | |
| XM_018092030.1 | XP_017947519.1 | 5 - 6 | 664 | 84 | ||
| GenBank mRNA | BC075530.1 | - | 6 - 7 | 699 | 84 | |
| CR762186.2 | - | 6 - 7 | 647 | 84 | ||
More Information
Set Membership: |
|

yeats4
Genomic Map