Assay Details
| Gene Symbol: gadd45g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004986.1 | NP_001004986.1 | 3 - 4 | 471 | 69 | |
| GenBank mRNA | BC075545.1 | - | 3 - 4 | 471 | 69 | |
| CR761710.2 | - | 3 - 4 | 466 | 69 | ||
More Information
Set Membership: |
|

gadd45g
Genomic Map