Assay Details
| Gene Symbol: yars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004987.1 | NP_001004987.1 | 1453 | 72 | ||
| GenBank mRNA | BC075547.1 | - | 1453 | 72 | ||
| CT485781.1 | - | 1414 | 72 | |||
More Information
Set Membership: |
|

yars
Genomic Map