Assay Details
| Gene Symbol: skap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004990.1 | NP_001004990.1 | 2 - 3 | 193 | 64 | |
| XM_012971641.1 | XP_012827095.1 | 2 - 3 | 242 | 64 | ||
| GenBank mRNA | BC075553.1 | - | 2 - 3 | 193 | 64 | |
More Information
Set Membership: |
|

skap1
Genomic Map