Assay Details
| Gene Symbol: riok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004996.1 | NP_001004996.1 | 1 - 2 | 334 | 57 | |
| XM_012964531.1 | XP_012819985.1 | 1 - 2 | 379 | 57 | ||
| GenBank mRNA | BC075585.1 | - | 1 - 2 | 334 | 57 | |
| CR760584.2 | - | 1 - 2 | 225 | 57 | ||
More Information
Set Membership: |
|

riok3
Genomic Map