Assay Details
| Gene Symbol: trarg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005003.1 | NP_001005003.1 | 1 - 2 | 621 | 74 | |
| GenBank mRNA | BC076649.1 | - | 1 - 2 | 621 | 74 | |
More Information
Set Membership: |
|

trarg1
Genomic Map