Assay Details
| Gene Symbol: trarg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005003.1 | NP_001005003.1 | 3 | 2119 | 106 | |
| GenBank mRNA | BC076649.1 | - | 3 | 2119 | 106 | |
More Information
Set Membership: |
|

trarg1
Genomic Map