Assay Details
| Gene Symbol: tmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005010.1 | NP_001005010.1 | 1 - 2 | 300 | 69 | |
| XM_012966413.2 | XP_012821867.1 | 1 - 2 | 333 | 69 | ||
| XM_018095349.1 | XP_017950838.1 | 1 - 2 | 333 | 69 | ||
| GenBank mRNA | BC076663.1 | - | 1 - 2 | 300 | 69 | |
| CR761011.2 | - | 1 - 2 | 297 | 69 | ||
More Information
Set Membership: |
|

tmx2
Genomic Map