Assay Details
| Gene Symbol: rpl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005020.1 | NP_001005020.1 | 5 - 6 | 537 | 61 | |
| GenBank mRNA | BC076695.1 | - | 5 - 6 | 537 | 61 | |
| CU075449.1 | - | 4 - 5 | 531 | 61 | ||
More Information
Set Membership: |
|

rpl7
Genomic Map