Assay Details
| Gene Symbol: tomm20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005024.1 | NP_001005024.1 | 1 - 2 | 216 | 72 | |
| GenBank mRNA | BC076705.1 | - | 1 - 2 | 216 | 72 | |
| CR760251.2 | - | 1 - 2 | 151 | 72 | ||
More Information
Set Membership: |
|

tomm20
Genomic Map