Assay Details
| Gene Symbol: tpm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005028.1 | NP_001005028.1 | 1 - 2 | 361 | 68 | |
| XM_012966419.1 | XP_012821873.1 | 1 - 2 | 458 | 68 | ||
| XM_012966420.1 | XP_012821874.1 | 1 - 2 | 458 | 68 | ||
| XM_012966421.2 | XP_012821875.1 | 1 - 2 | 458 | 68 | ||
| XM_018092035.1 | XP_017947524.1 | 1 - 2 | 458 | 68 | ||
| GenBank mRNA | BC076711.1 | - | 1 - 2 | 361 | 68 | |
| CT010525.2 | - | 1 - 2 | 167 | 68 | ||
More Information
Set Membership: |
|

tpm1
Genomic Map