Assay Details
| Gene Symbol: spc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005041.1 | NP_001005041.1 | 3 - 4 | 347 | 105 | |
| XM_012968241.2 | XP_012823695.1 | 3 - 4 | 325 | 105 | ||
| XM_012968243.2 | XP_012823697.1 | 3 - 4 | 373 | 105 | ||
| XM_012968244.2 | XP_012823698.1 | 3 - 4 | 363 | 105 | ||
| XM_012968245.2 | XP_012823699.1 | 3 - 4 | 332 | 105 | ||
| XM_012968246.2 | XP_012823700.1 | 3 - 4 | 257 | 105 | ||
| GenBank mRNA | BC076904.1 | - | 3 - 4 | 347 | 105 | |
| CR761196.2 | - | 2 - 3 | 238 | 105 | ||
More Information
Set Membership: |
|

spc25
Genomic Map