Assay Details
| Gene Symbol: chmp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005047.1 | NP_001005047.1 | 5 - 6 | 523 | 99 | |
| GenBank mRNA | BC076916.1 | - | 5 - 6 | 523 | 99 | |
| CR760798.2 | - | 5 - 6 | 505 | 99 | ||
More Information
Set Membership: |
|

chmp1b
Genomic Map