Assay Details
| Gene Symbol: pabpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005051.1 | NP_001005051.1 | 9 - 10 | 1611 | 62 | |
| GenBank mRNA | BC076931.1 | - | 9 - 10 | 1611 | 62 | |
| CT025226.2 | - | 9 - 10 | 1624 | 62 | ||
More Information
Set Membership: |
|

pabpc1
Genomic Map