Assay Details
| Gene Symbol: psmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005073.1 | NP_001005073.1 | 5 - 6 | 582 | 80 | |
| XM_012968255.2 | XP_012823709.1 | 5 - 6 | 699 | 80 | ||
| GenBank mRNA | BC076990.1 | - | 5 - 6 | 582 | 80 | |
| CU075836.1 | - | 5 - 6 | 569 | 80 | ||
More Information
Set Membership: |
|

psmd8
Genomic Map