Assay Details
| Gene Symbol: vps29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005079.2 | NP_001005079.1 | 3 - 4 | 468 | 73 | |
| XM_012960142.2 | XP_012815596.1 | 4 - 5 | 557 | 73 | ||
| GenBank mRNA | BC077001.1 | - | 3 - 4 | 470 | 73 | |
| CR761433.2 | - | 2 - 3 | 435 | 73 | ||
More Information
Set Membership: |
|

vps29
Genomic Map