Assay Details
| Gene Symbol: hpgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005090.1 | NP_001005090.1 | 2 - 3 | 197 | 71 | |
| XM_012960165.2 | XP_012815619.1 | 3 - 4 | 255 | 71 | ||
| GenBank mRNA | BC077016.1 | - | 2 - 3 | 197 | 71 | |
| BC166947.1 | - | 2 - 3 | 158 | 71 | ||
| CR761642.2 | - | 2 - 3 | 168 | 71 | ||
More Information
Set Membership: |
|

hpgds
Genomic Map