Assay Details
| Gene Symbol: rpl36a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005095.1 | NP_001005095.1 | 3 - 4 | 205 | 68 | |
| GenBank mRNA | BC077026.1 | - | 3 - 4 | 205 | 68 | |
| CR926212.1 | - | 2 - 3 | 183 | 68 | ||
| CR942739.2 | - | 194 | 68 | |||
More Information
Set Membership: |
|

rpl36a
Genomic Map