Assay Details
| Gene Symbol: cyp2a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010998.1 | NP_001010998.1 | 4 - 5 | 655 | 72 | |
| GenBank mRNA | BC075418.1 | - | 4 - 5 | 655 | 72 | |
More Information
Set Membership: |
|

cyp2a6
Genomic Map