Assay Details
| Gene Symbol: mfap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011003.1 | NP_001011003.1 | 4 - 5 | 722 | 90 | |
| GenBank mRNA | BC082615.1 | - | 4 - 5 | 722 | 90 | |
| CR762333.2 | - | 4 - 5 | 663 | 90 | ||
More Information
Set Membership: |
|

mfap1
Genomic Map