Assay Details
| Gene Symbol: rph3al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011007.1 | NP_001011007.1 | 7 - 8 | 800 | 63 | |
| XM_012956827.2 | XP_012812281.1 | 8 - 9 | 1149 | 63 | ||
| XM_012956828.2 | XP_012812282.1 | 8 - 9 | 1117 | 63 | ||
| XM_012956829.2 | XP_012812283.1 | 8 - 9 | 1049 | 63 | ||
| XM_012956830.2 | XP_012812284.1 | 8 - 9 | 1017 | 63 | ||
| XM_012956832.2 | XP_012812286.1 | 7 - 8 | 985 | 63 | ||
| XM_012956833.2 | XP_012812287.1 | 7 - 8 | 954 | 63 | ||
| XM_012956834.2 | XP_012812288.1 | 8 - 9 | 1147 | 63 | ||
| XM_012956835.2 | XP_012812289.1 | 8 - 9 | 1143 | 63 | ||
| XM_012956836.2 | XP_012812290.1 | 8 - 9 | 1043 | 63 | ||
| GenBank mRNA | BC082720.1 | - | 7 - 8 | 800 | 63 | |
| CR761174.2 | - | 8 - 9 | 857 | 63 | ||
More Information
Set Membership: |
|

rph3al
Genomic Map