Assay Details
| Gene Symbol: c1orf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018091199.1 | XP_017946688.1 | 2 - 3 | 951 | 110 | |
| GenBank mRNA | BC082737.1 | - | 2 - 3 | 955 | 110 | |
More Information
Set Membership: |
|

c1orf74
Genomic Map