Assay Details
| Gene Symbol: gng13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011028.1 | NP_001011028.1 | 3 - 4 | 275 | 68 | |
| XM_012970353.2 | XP_012825807.1 | 4 - 5 | 708 | 68 | ||
| XM_012970354.2 | XP_012825808.1 | 3 - 4 | 357 | 68 | ||
| XM_018097093.1 | XP_017952582.1 | 2 - 3 | 467 | 68 | ||
| GenBank mRNA | BC082958.1 | - | 3 - 4 | 275 | 68 | |
| CR760269.2 | - | 3 - 4 | 272 | 68 | ||
More Information
Set Membership: |
|

gng13
Genomic Map