Assay Details
| Gene Symbol: rpl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011030.2 | NP_001011030.1 | 3 - 4 | 274 | 92 | |
| GenBank mRNA | BC082960.1 | - | 3 - 4 | 275 | 92 | |
| CR760488.2 | - | 3 - 4 | 213 | 92 | ||
More Information
Set Membership: |
|

rpl18
Genomic Map