Assay Details
| Gene Symbol: slc35a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011040.1 | NP_001011040.1 | 4 - 5 | 539 | 64 | |
| XM_012962930.2 | XP_012818384.1 | 4 - 5 | 657 | 64 | ||
| GenBank mRNA | BC084147.1 | - | 4 - 5 | 539 | 64 | |
| CT485743.1 | - | 4 - 5 | 546 | 64 | ||
More Information
Set Membership: |
|

slc35a1
Genomic Map